Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286272.1 | NP_001273201.1 | 6 | 1008 | ||
NM_001286273.1 | NP_001273202.1 | 5 | 769 | |||
NM_003295.3 | NP_003286.1 | 6 | 843 | |||
GenBank mRNA | AK130847.1 | - | 6 | 742 | ||
AK296587.1 | - | 6 | 652 | |||
AY117678.1 | - | 6 | 743 | |||
BC003352.2 | - | 6 | 726 | 2899964 | ||
BC012431.1 | - | 6 | 702 | 3880559 | ||
BC022436.1 | - | 5 | 697 | 4281364 | ||
BC040008.1 | - | 3 | 1749 | |||
BC052333.1 | - | 6 | 732 | 6058799 | ||
L13806.1 | - | 4 | 463 | |||
X16064.1 | - | 6 | 722 | |||
X64899.1 | - | 6 | 714 |