Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPTE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290224.1 | NP_001277153.1 | 15, 16 | 1399 | ||
NM_199259.3 | NP_954868.1 | 19, 20 | 1621 | |||
NM_199260.3 | NP_954869.1 | 18, 19 | 1561 | |||
NM_199261.3 | NP_954870.2 | 20, 21 | 1675 | |||
GenBank mRNA | AF007118.1 | - | 20, 21 | 1623 | ||
AF495908.1 | - | 19, 20 | 1569 | |||
AK308254.1 | - | 19, 20 | 1518 | |||
AK314285.1 | - | 20, 21 | 1574 | |||
AL833345.1 | - | 20, 21 | 1582 | |||
AY219886.1 | - | 19, 20 | 1569 | |||
AY219887.1 | - | 18, 19 | 1509 | |||
AY219888.1 | - | 15, 16 | 1347 | |||
BC028719.1 | - | 19, 20 | 1604 | 5269598 |