Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmp17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105925.1 | NP_001099395.1 | 9 | 1464 | ||
XM_008769195.1 | XP_008767417.1 | 9 | 2747 |