Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRISP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142407.2 | NP_001135879.1 | 9 | 835 | ||
NM_001142408.2 | NP_001135880.1 | 10 | 892 | |||
NM_001142417.2 | NP_001135889.1 | 10 | 872 | |||
NM_001142435.2 | NP_001135907.1 | 9 | 855 | |||
NM_001261822.1 | NP_001248751.1 | 9 | 861 | |||
NM_003296.3 | NP_003287.1 | 10 | 898 | |||
XM_005249349.1 | XP_005249406.1 | 11 | 977 | |||
XM_005249350.1 | XP_005249407.1 | 10 | 935 | |||
XM_005249351.3 | XP_005249408.1 | 10 | 955 | |||
XM_005249352.3 | XP_005249409.1 | 10 | 946 | |||
XM_005249353.3 | XP_005249410.1 | 11 | 982 | |||
XM_005249356.1 | XP_005249413.1 | 11 | 980 | |||
XM_011514841.1 | XP_011513143.1 | 11 | 1312 | |||
XM_011514842.1 | XP_011513144.1 | 10 | 1275 | |||
GenBank mRNA | AK128009.1 | - | 1 | 1828 | ||
AK223337.1 | - | 10 | 835 | |||
AK292385.1 | - | 10 | 876 | |||
AY289796.1 | - | 10 | 842 | |||
AY292862.1 | - | 10 | 937 | |||
AY292863.1 | - | 9 | 836 | |||
AY773479.1 | - | 10 | 878 | |||
BC022011.2 | - | 10 | 845 | 4826427 | ||
BC107707.1 | - | 9 | 792 | 6618679 | ||
BI831679.1 | - | 4 | 247 | |||
BM552843.1 | - | 9 | 770 | |||
BP369727.1 | - | 4 | 349 | |||
M25532.1 | - | 10 | 821 | |||
X95239.1 | - | 10 | 872 |