Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSP90B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003299.2 | NP_003290.1 | 2 | 244 | ||
GenBank mRNA | AB209534.1 | - | 2 | 140 | ||
AJ890084.1 | - | 2 | 196 | |||
AK025459.1 | - | 2 | 196 | |||
AK298996.1 | - | 2 | 168 | |||
AK300521.1 | - | 2 | 167 | |||
BC009195.2 | - | 2 | 140 | |||
BC066656.1 | - | 2 | 158 | 6165138 | ||
EU794608.1 | - | 2 | 158 | |||
FJ460515.1 | - | 2 | 158 | |||
X15187.1 | - | 2 | 167 |