Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sgsm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105937.2 | NP_001099407.2 | 20 | 2712 | ||
XM_008769322.1 | XP_008767544.1 | 21 | 3216 |