Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079936.1 | NP_001073405.1 | 2 | 410 | ||
XM_006240750.2 | XP_006240812.1 | 2 | 333 | |||
XM_006240751.2 | XP_006240813.1 | 3 | 431 | |||
XM_006240752.2 | XP_006240814.1 | 2 | 558 | |||
GenBank mRNA | BC129101.1 | - | 2 | 410 | 7098463 | |
FQ211489.1 | - | 2 | 548 |