Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr908 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001360.1 | NP_001001360.1 | 1 | 269 |
Gene Symbol: Olr1845 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000696.1 | NP_001000696.1 | 1 | 269 | ||
XM_006240816.1 | XP_006240878.1 | 1 | 269 |
Gene Symbol: Olr916 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000587.1 | NP_001000587.1 | 1 | 269 |
Gene Symbol: Olr991 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008765073.1 | XP_008763295.1 | 1 | 110 |