Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGD1564093 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105946.1 | NP_001099416.1 | 2 | 430 | ||
GenBank mRNA | FQ231178.1 | - | 2 | 369 | ||
FQ235293.1 | - | 2 | 369 |
Gene Symbol: LOC501934 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773502.1 | XP_008771724.1 | 2 | 106 |