Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pik3c2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105951.1 | NP_001099421.1 | 24 | 4234 | ||
XM_006249754.2 | XP_006249816.1 | 25 | 4337 | |||
XM_006249756.2 | XP_006249818.1 | 24 | 3780 | |||
XM_008769442.1 | XP_008767664.1 | 25 | 5177 |