Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem183a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013871.1 | NP_001013893.1 | 7 | 875 | ||
XM_006249849.2 | XP_006249911.1 | 7 | 939 | |||
XM_006249850.2 | XP_006249912.1 | 6 | 863 | |||
GenBank mRNA | BC079375.1 | - | 7 | 875 | 7110239 | |
FQ210515.1 | - | 7 | 908 |