Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adipor1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207587.1 | NP_997470.1 | 6 | 856 | ||
XM_006249852.2 | XP_006249914.1 | 6 | 892 | |||
XM_008769543.1 | XP_008767765.1 | 6 | 1193 | |||
XM_008769544.1 | XP_008767766.1 | 6 | 1555 | |||
GenBank mRNA | BC061838.1 | - | 6 | 856 | 5599938 | |
DQ148391.1 | - | 5 | 644 |