Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003302.2 | NP_003293.2 | 3 | 464 | ||
GenBank mRNA | AB628085.1 | - | 3 | 231 | ||
AB628086.1 | - | 3 | 244 | |||
AB628087.1 | - | 2 | 166 | |||
AB628088.1 | - | 3 | 294 | |||
AF000974.1 | - | 3 | 453 | |||
AJ001902.1 | - | 3 | 366 | |||
AK056773.1 | - | 3 | 358 | |||
AK291906.1 | - | 3 | 401 | |||
AK309312.1 | - | 3 | 355 | |||
BC002680.2 | - | 3 | 357 | 3610413 | ||
BC004249.1 | - | 3 | 369 | 3621877 | ||
BC004999.1 | - | 3 | 363 | 2958142 | ||
BC021540.1 | - | 3 | 369 | 3622017 | ||
BC028985.1 | - | 3 | 376 | 5122169 | ||
BM800083.1 | - | 3 | 464 | |||
CR749505.1 | - | 3 | 292 |