Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003302.2 | NP_003293.2 | 8 | 1422 | ||
GenBank mRNA | AB628085.1 | - | 8 | 1189 | ||
AB628086.1 | - | 8 | 1202 | |||
AB628087.1 | - | 7 | 1124 | |||
AB628088.1 | - | 8 | 1005 | |||
AF000974.1 | - | 8 | 1411 | |||
AF025437.1 | - | 5 | 595 | |||
AJ001902.1 | - | 8 | 1324 | |||
AK056773.1 | - | 7 | 1426 | |||
AK291906.1 | - | 8 | 1359 | |||
BC002680.2 | - | 8 | 1315 | 3610413 | ||
BC004249.1 | - | 8 | 1327 | 3621877 | ||
BC004999.1 | - | 8 | 1321 | 2958142 | ||
BC021540.1 | - | 8 | 1327 | 3622017 | ||
BC028985.1 | - | 8 | 1334 | 5122169 | ||
CR749505.1 | - | 7 | 2049 | |||
L40374.1 | - | 3 | 325 |