Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rgs18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047084.1 | NP_001040549.1 | 3 | 257 | ||
XM_008769548.1 | XP_008767770.1 | 3 | 283 | |||
GenBank mRNA | AY651776.1 | - | 3 | 257 |
Gene Symbol: LOC103690097 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008769583.1 | XP_008767805.1 | 506 |