Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039705.2 | NP_001034794.1 | 3 | 1150 | ||
NM_016157.3 | NP_057241.2 | 3 | 1150 | |||
NM_177556.2 | NP_808224.1 | 3 | 1135 | |||
NR_073148.1 | 3 | 1135 | ||||
XM_006724600.2 | XP_006724663.1 | 3 | 916 | |||
XM_011530808.1 | XP_011529110.1 | 3 | 961 | |||
XM_011530809.1 | XP_011529111.1 | 3 | 912 | |||
XM_011530811.2 | XP_011529113.1 | 3 | 1298 | |||
XM_011530812.1 | XP_011529114.1 | 3 | 899 | |||
XM_011530813.1 | XP_011529115.1 | 3 | 949 | |||
XM_011530814.1 | XP_011529116.1 | 3 | 843 | |||
XM_017029767.1 | XP_016885256.1 | 3 | 1313 | |||
XM_017029768.1 | XP_016885257.1 | 3 | 930 | |||
XM_017029769.1 | XP_016885258.1 | 3 | 912 | |||
XM_017029770.1 | XP_016885259.1 | 3 | 961 | |||
XM_017029771.1 | XP_016885260.1 | 3 | 1289 | |||
XM_017029772.1 | XP_016885261.1 | 3 | 930 | |||
XM_017029773.1 | XP_016885262.1 | 3 | 841 | |||
GenBank mRNA | AB029037.2 | - | 3 | 871 | ||
AF349719.1 | - | 3 | 882 | |||
AF349720.1 | - | 3 | 882 | |||
AK001300.1 | - | 1 | 580 | |||
AK027489.1 | - | 3 | 889 | |||
AK299568.1 | - | 2626 | ||||
BC026914.1 | - | 3 | 865 |