Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006250119.2 | XP_006250181.1 | 2 | 551 | ||
XM_006250122.2 | XP_006250184.1 | 2 | 546 | |||
XM_006250123.2 | XP_006250185.1 | 2 | 542 | |||
XM_008769703.1 | XP_008767925.1 | 4 | 607 | |||
XM_008769704.1 | XP_008767926.1 | 2 | 543 |