Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dpt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105965.1 | NP_001099435.1 | 2 | 464 | ||
XM_006250153.1 | XP_006250215.1 | 2 | 701 | |||
XM_006250154.1 | XP_006250216.1 | 2 | 701 | |||
XM_008769624.1 | XP_008767846.1 | 2 | 701 | |||
GenBank mRNA | BC167054.1 | - | 2 | 459 | 7113939 |