Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr161 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751260.3 | XP_003751308.1 | 3 | 1052 | ||
XM_006250191.2 | XP_006250253.1 | 3 | 1122 | |||
XM_008769710.1 | XP_008767932.1 | 4 | 1133 | |||
XM_008769711.1 | XP_008767933.1 | 3 | 1071 | |||
XM_008769712.1 | XP_008767934.1 | 3 | 1050 |
Set Membership: |
G-protein coupled receptor |