Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dcaf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006250194.2 | XP_006250256.1 | 4, 5 | 744 | ||
XM_006250195.2 | XP_006250257.1 | 4, 5 | 744 | |||
XM_006250196.2 | XP_006250258.1 | 4, 5 | 744 | |||
XM_006250197.2 | XP_006250259.1 | 4, 5 | 744 | |||
XM_006250198.2 | XP_006250260.1 | 4, 5 | 744 | |||
XM_006250199.2 | XP_006250261.1 | 4, 5 | 744 | |||
XM_006250200.2 | XP_006250262.1 | 4, 5 | 744 | |||
XM_006250201.2 | XP_006250263.1 | 4, 5 | 744 | |||
XM_213926.8 | XP_213926.5 | 4, 5 | 744 |