Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nhlh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105970.1 | NP_001099440.1 | 2 | 1137 | ||
XM_006250280.1 | XP_006250342.1 | 3 | 1229 |