Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Disp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105983.1 | NP_001099453.1 | 9 | 1385 | ||
XM_006250381.2 | XP_006250443.1 | 8 | 1254 | |||
XM_006250383.2 | XP_006250445.1 | 7 | 1715 | |||
XM_006250384.2 | XP_006250446.1 | 7 | 1194 | |||
XM_008769827.1 | XP_008768049.1 | 10 | 2079 | |||
XM_008769828.1 | XP_008768050.1 | 8 | 1365 |