Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001261401.1 | NP_001248330.1 | 3, 4 | 484 | ||
NM_004622.2 | NP_004613.1 | 3, 4 | 484 | |||
NR_048556.1 | 2, 3 | 390 | ||||
GenBank mRNA | AK091915.1 | - | 3, 4 | 467 | ||
AK222868.1 | - | 3, 4 | 330 | |||
AK296469.1 | - | 3, 4 | 348 | |||
AK298812.1 | - | 2, 3 | 243 | |||
AK307344.1 | - | 3, 4 | 319 | |||
AK315763.1 | - | 3, 4 | 369 | |||
BC002359.1 | - | 3, 4 | 363 | 2822564 | ||
BT019490.1 | - | 3, 4 | 249 | |||
BT019491.1 | - | 3, 4 | 249 | |||
DA523145.1 | - | 2, 3 | 273 | |||
DA772798.1 | - | 3, 4 | 485 | |||
X78627.1 | - | 3, 4 | 330 |