Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001261401.1 | NP_001248330.1 | 5 | 753 | ||
NM_004622.2 | NP_004613.1 | 6 | 833 | |||
NR_048556.1 | 5 | 739 | ||||
NR_048557.1 | 4 | 642 | ||||
NR_048558.1 | 3 | 526 | ||||
NR_048559.1 | 4 | 750 | ||||
GenBank mRNA | AK091915.1 | - | 6 | 816 | ||
AK222868.1 | - | 6 | 679 | |||
AK296469.1 | - | 5 | 617 | |||
AK298812.1 | - | 5 | 592 | |||
AK307344.1 | - | 6 | 668 | |||
AK311410.1 | - | 3 | 411 | |||
AK315763.1 | - | 6 | 718 | |||
BC002359.1 | - | 6 | 712 | 2822564 | ||
BP289175.1 | - | 4 | 527 | |||
BT019490.1 | - | 6 | 598 | |||
BT019491.1 | - | 6 | 598 | |||
X78627.1 | - | 6 | 679 |