Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001261401.1 | NP_001248330.1 | 2 | 354 | ||
NM_004622.2 | NP_004613.1 | 2 | 354 | |||
GenBank mRNA | AK091915.1 | - | 2 | 236 | ||
AK222868.1 | - | 2 | 200 | |||
AK296469.1 | - | 2 | 218 | |||
AK307344.1 | - | 2 | 196 | |||
AK315763.1 | - | 2 | 239 | |||
BC002359.1 | - | 2 | 233 | 2822564 | ||
BT019490.1 | - | 2 | 119 | |||
BT019491.1 | - | 2 | 119 | |||
DA772798.1 | - | 2 | 355 | |||
X78627.1 | - | 2 | 200 |