Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif21b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105990.1 | NP_001099460.1 | 25 | 3806 | ||
XM_006249853.2 | XP_006249915.1 | 25 | 3853 | |||
XM_006249854.2 | XP_006249916.1 | 25 | 3853 | |||
XM_006249855.2 | XP_006249917.1 | 25 | 3850 | |||
XM_006249856.2 | XP_006249918.1 | 25 | 3853 | |||
XM_006249857.2 | XP_006249919.1 | 25 | 3850 | |||
XM_006249858.2 | XP_006249920.1 | 25 | 3849 |