Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glmn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105993.1 | NP_001099463.1 | 7 | 786 | ||
XM_008769921.1 | XP_008768143.1 | 6 | 1393 | |||
XM_008769922.1 | XP_008768144.1 | 7 | 787 | |||
XM_008769923.1 | XP_008768145.1 | 7 | 866 | |||
XM_008769924.1 | XP_008768146.1 | 6 | 1393 | |||
XM_008769925.1 | XP_008768147.1 | 5 | 1452 | |||
XM_008769926.1 | XP_008768148.1 | 5 | 579 | |||
GenBank mRNA | BC083633.1 | - | 6 | 726 | ||
BC100158.1 | - | 7 | 899 | |||
BC162027.1 | - | 7 | 762 |