Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105995.1 | NP_001099465.1 | 4 | 1170 | ||
XM_008770004.1 | XP_008768226.1 | 6 | 1505 | |||
XM_008770005.1 | XP_008768227.1 | 6 | 1409 | |||
XM_008770006.1 | XP_008768228.1 | 6 | 1347 | |||
XM_008770007.1 | XP_008768229.1 | 5 | 1174 |