Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Naa11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024742.1 | NP_001019913.1 | 1 | 432 | ||
XM_006250697.2 | XP_006250759.1 | 1 | 453 | |||
GenBank mRNA | BC078912.1 | - | 1 | 405 | ||
BC097350.1 | - | 1 | 432 | 7462336 |