Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105999.1 | NP_001099469.1 | 27 | 7058 | ||
XM_006250763.2 | XP_006250825.1 | 29 | 6865 | |||
XM_006250764.2 | XP_006250826.1 | 29 | 6862 | |||
XM_006250765.2 | XP_006250827.1 | 29 | 6811 | |||
XM_006250766.2 | XP_006250828.1 | 29 | 7447 | |||
XM_006250767.2 | XP_006250829.1 | 29 | 7747 | |||
XM_006250768.2 | XP_006250830.1 | 28 | 6037 | |||
XM_006250769.2 | XP_006250831.1 | 28 | 6034 | |||
XM_006250770.2 | XP_006250832.1 | 28 | 5983 | |||
XM_008770011.1 | XP_008768233.1 | 29 | 7444 |