Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ociad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013874.1 | NP_001013896.1 | 4, 5 | 347 | ||
GenBank mRNA | BC083718.1 | - | 4, 5 | 347 | 7190773 | |
FQ212940.1 | - | 4, 5 | 312 | |||
FQ213030.1 | - | 4, 5 | 356 | |||
FQ213629.1 | - | 4, 5 | 384 |