Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSNAX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005999.2 | NP_005990.1 | 6 | 870 | ||
GenBank mRNA | AK313068.1 | - | 6 | 808 | ||
BC010376.1 | - | 6 | 828 | 4283594 | ||
BC011797.1 | - | 6 | 845 | 4138517 | ||
BX537610.1 | - | 5 | 3838 | |||
X95073.1 | - | 6 | 872 |