Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSSC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003310.2 | NP_003301.1 | 5 | 682 | ||
XM_006711893.2 | XP_006711956.1 | 3 | 587 | |||
XM_011510383.2 | XP_011508685.1 | 6 | 818 | |||
XM_011510384.2 | XP_011508686.1 | 6 | 711 | |||
XM_011510385.2 | XP_011508687.1 | 5 | 711 | |||
XM_011510387.2 | XP_011508689.1 | 4 | 473 | |||
XM_017004818.1 | XP_016860307.1 | 5 | 783 | |||
GenBank mRNA | AF019952.1 | - | 5 | 640 | ||
AK026522.1 | - | 5 | 623 | |||
AK126288.1 | - | 8 | 1671 | |||
AK222652.1 | - | 5 | 618 | |||
BC002485.2 | - | 5 | 590 | 3349430 |