Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Osm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006961.1 | NP_001006962.1 | 2, 3 | 174 | ||
GenBank mRNA | AB167521.1 | - | 2, 3 | 174 |