Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: H2afv | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106019.1 | NP_001099489.1 | 3 | 204 | ||
XM_006251387.2 | XP_006251449.1 | 3 | 542 |
Gene Symbol: RGD1560813 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751821.2 | XP_003751869.1 | 1 | 158 |