Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TST | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270483.1 | NP_001257412.1 | 2 | 527 | ||
NM_003312.5 | NP_003303.2 | 2 | 559 | |||
GenBank mRNA | AK000862.1 | - | 2 | 553 | ||
AK091874.1 | - | 1 | 1179 | |||
AK301300.1 | - | 2 | 553 | |||
BC010148.2 | - | 2 | 536 | 4329727 | ||
BQ231617.1 | - | 2 | 552 | |||
CR456598.1 | - | 1 | 562 | |||
CR456998.1 | - | 1 | 484 | |||
D87292.1 | - | 2 | 532 |