Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc104 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024866.1 | NP_001020037.1 | 1, 2 | 234 | ||
XM_006251598.2 | XP_006251660.1 | 1, 2 | 328 | |||
GenBank mRNA | BC097425.1 | - | 1, 2 | 234 | 7455112 |