Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmed5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007619.1 | NP_001007620.1 | 4 | 638 | ||
GenBank mRNA | AY539871.1 | - | 550 | |||
BC079452.1 | - | 4 | 638 | 7121068 |