Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mss51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106025.1 | NP_001099495.1 | 5 | 664 | ||
GenBank mRNA | FQ215928.1 | - | 3 | 311 |