Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282500.1 | NP_001269429.1 | 6, 7 | 818 | ||
NM_003314.2 | NP_003305.1 | 6, 7 | 821 | |||
GenBank mRNA | AK222641.1 | - | 6, 7 | 772 | ||
AK222984.1 | - | 6, 7 | 766 | |||
AK315262.1 | - | 6, 7 | 776 | |||
BC000942.3 | - | 5, 6 | 726 | 3446109 | ||
U46570.1 | - | 6, 7 | 748 |