Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr1611 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751458.3 | XP_003751506.1 | 1 | 137 |
Gene Symbol: Olr1610 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000094.1 | NP_001000094.1 | 1 | 99 | ||
XM_008770652.1 | XP_008768874.1 | 1 | 99 |