Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJC7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144766.2 | NP_001138238.1 | 12 | 1389 | ||
NM_003315.3 | NP_003306.3 | 12 | 1621 | |||
NR_029431.1 | 11 | 1532 | ||||
XM_011525167.2 | XP_011523469.1 | 13 | 1551 | |||
XM_011525168.2 | XP_011523470.1 | 13 | 1511 | |||
XM_011525169.2 | XP_011523471.1 | 12 | 1363 | |||
XM_017024994.1 | XP_016880483.1 | 13 | 1545 | |||
GenBank mRNA | AK291047.1 | - | 12 | 1322 | ||
AK296602.1 | - | 12 | 1382 | |||
AK298860.1 | - | 12 | 1337 | |||
AK316081.1 | - | 11 | 1258 | |||
BC003601.1 | - | 11 | 1244 | 3543074 | ||
BC011837.2 | - | 12 | 1340 | 3945721 | ||
BC033772.1 | - | 12 | 1366 | 3924081 | ||
BX647209.1 | - | 12 | 1577 | |||
U46571.1 | - | 12 | 1336 |