Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pnp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106031.1 | NP_001099501.1 | 5 | 567 | ||
XM_006251857.2 | XP_006251919.1 | 5 | 597 | |||
GenBank mRNA | FQ209687.1 | - | 2 | 121 | ||
FQ210355.1 | - | 2 | 121 | |||
FQ213258.1 | - | 2 | 121 | |||
FQ213367.1 | - | 2 | 121 | |||
FQ214084.1 | - | 2 | 121 | |||
FQ219529.1 | - | 2 | 121 | |||
FQ225420.1 | - | 2 | 121 | |||
FQ225955.1 | - | 2 | 121 | |||
FQ228749.1 | - | 2 | 121 |