Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC685078 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | EX488681.1 | - | 732 |
Gene Symbol: LOC290071 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY227970.1 | - | 5, 6 | 718 | ||
BC088274.1 | - | 5, 6 | 740 | |||
BC088276.1 | - | 5, 6 | 722 | 7381661 | ||
BC091200.1 | - | 5, 6 | 774 | |||
BC091267.1 | - | 6, 7 | 707 | 7380098 | ||
BC091273.1 | - | 5, 6 | 884 | 7383056 | ||
BC092593.1 | - | 5, 6 | 754 | 7379916 | ||
BC092595.1 | - | 5, 6 | 857 | 7382388 | ||
BC092596.1 | - | 5, 6 | 722 | 7382760 | ||
BC092597.1 | - | 5, 6 | 711 | |||
BC093389.1 | - | 5, 6 | 708 | 7387331 | ||
BC093392.1 | - | 5, 6 | 759 | 7388480 | ||
DQ340291.1 | - | 5, 6 | 706 | |||
M18853.1 | - | 5, 6 | 687 |