Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mphosph8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017375.2 | NP_001017375.2 | 3 | 1002 | ||
GenBank mRNA | BC099116.1 | - | 3 | 945 | 7462184 |