Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: N6amt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134990.1 | NP_001128462.1 | 4 | 487 | ||
XM_006252059.2 | XP_006252121.1 | 5 | 1340 | |||
XM_006252061.2 | XP_006252123.1 | 4 | 1168 | |||
GenBank mRNA | BC168660.1 | - | 4 | 487 | 5621479 | |
FQ230419.1 | - | 5 | 537 |