Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xpo4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106042.1 | NP_001099512.1 | 15, 16 | 2345 | ||
XM_006252062.2 | XP_006252124.1 | 15, 16 | 2349 | |||
XM_006252063.2 | XP_006252125.1 | 15, 16 | 2221 | |||
GenBank mRNA | FQ225683.1 | - | 3, 4 | 418 | ||
FQ231595.1 | - | 3, 4 | 418 | |||
FQ234166.1 | - | 3, 4 | 418 |