Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000370.3 | NP_000361.1 | 3 | 456 | ||
GenBank mRNA | BC041784.1 | - | 3 | 442 | ||
BC058000.1 | - | 3 | 460 | 4610804 | ||
D49488.1 | - | 3 | 442 | |||
U21938.1 | - | 3 | 431 |