Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dok2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106048.1 | NP_001099518.1 | 3, 4 | 557 | ||
XM_006252262.1 | XP_006252324.1 | 4, 5 | 1326 | |||
XM_006252264.2 | XP_006252326.1 | 4, 5 | 1787 | |||
XM_008770826.1 | XP_008769048.1 | 3, 4 | 2761 |