Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000371.3 | NP_000362.1 | 4 | 645 | ||
GenBank mRNA | BC005310.1 | - | 4 | 535 | 4071761 | |
BC020791.1 | - | 4 | 535 | 4734010 | ||
CA948224.1 | - | 1 | 28 | |||
D00096.1 | - | 4 | 535 | |||
GU727633.1 | - | 4 | 535 | |||
K02091.1 | - | 4 | 535 | |||
M10605.1 | - | 4 | 534 | |||
M11714.1 | - | 3 | 421 |